Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 4 | 729 | ||
NM_023210.4 | NP_075699.3 | 4 | 729 | |||
GenBank mRNA | AB037685.1 | - | 4 | 404 | ||
AK012759.1 | - | 4 | 696 | |||
AK049647.1 | - | 4 | 696 | |||
AK076049.1 | - | 4 | 556 | |||
AK088401.1 | - | 4 | 709 | |||
AK147888.1 | - | 4 | 695 | |||
AK159446.1 | - | 4 | 696 | |||
AK168380.1 | - | 4 | 678 | |||
AK172146.1 | - | 4 | 743 | |||
AK193570.1 | - | 2 | 191 | |||
AK195817.1 | - | 2 | 100 | |||
AK199406.1 | - | 2 | 191 | |||
AK204175.1 | - | 2 | 191 | |||
BC005690.1 | - | 4 | 664 | 3596597 | ||
BC080684.1 | - | 4 | 669 | 6493476 | ||
CF952181.1 | - | 4 | 400 | |||
U89345.2 | - | 4 | 690 |