Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025616.3 | NP_079892.1 | 8 | 623 | ||
GenBank mRNA | AK008340.1 | - | 8 | 624 | ||
AK008382.1 | - | 8 | 621 | |||
AK010832.1 | - | 8 | 589 | |||
AK012977.1 | - | 8 | 620 | |||
AK088074.1 | - | 8 | 616 | |||
AK167473.1 | - | 8 | 626 | |||
AK179970.1 | - | 3 | 145 | |||
AK186280.1 | - | 3 | 145 | |||
AY551342.1 | - | 8 | 609 | |||
BC010303.1 | - | 8 | 598 | 3597676 | ||
BC025844.1 | - | 2 | 104 | |||
BC037056.1 | - | 8 | 625 |