Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175109.3 | NP_780318.1 | 2, 3 | 212 | ||
GenBank mRNA | AB233218.1 | - | 2, 3 | 208 | ||
AK010954.1 | - | 2, 3 | 196 | |||
AK075974.1 | - | 2, 3 | 193 | |||
BC038691.1 | - | 2, 3 | 198 | 1495043 | ||
BC050115.1 | - | 2, 3 | 201 | 30017673 | ||
BC065381.1 | - | 1, 2 | 145 | |||
BY237968.1 | - | 2, 3 | 212 |