Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023239.4 | NP_075728.1 | 1 | 453 | ||
GenBank mRNA | AF319979.1 | - | 1 | 331 | ||
AK010294.1 | - | 1 | 414 | |||
AK017727.1 | - | 1 | 455 | |||
AK018250.1 | - | 1 | 414 | |||
AK049759.1 | - | 1 | 433 | |||
AK076471.1 | - | 1 | 439 | |||
AK131157.1 | - | 410 | ||||
BC034892.1 | - | 1 | 378 | 1494952 | ||
BC092289.1 | - | 1 | 415 | 4990134 |