Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004361.2 | NP_001004361.2 | 4 | 549 | ||
NM_001142697.1 | NP_001136169.1 | 4 | 295 | |||
NM_001142698.1 | NP_001136170.1 | 4 | 549 | |||
GenBank mRNA | AK017728.1 | - | 4 | 303 | ||
AK160220.1 | - | 4 | 345 | |||
BC060253.1 | - | 2 | 101 | |||
BC080777.1 | - | 4 | 305 | 30439612 | ||
BQ770487.1 | - | 4 | 549 | |||
DV655125.1 | - | 4 | 295 |