Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025696.3 | NP_079972.1 | 12 | 1900 | ||
GenBank mRNA | AF276314.1 | - | 12 | 1920 | ||
AK158805.1 | - | 12 | 1767 |