Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025745.3 | NP_080021.3 | 7 | 684 | ||
GenBank mRNA | AK002471.1 | - | 7 | 675 | ||
AK077120.1 | - | 7 | 677 | |||
AK133874.1 | - | 7 | 685 | |||
BC018468.1 | - | 7 | 694 | 3591901 |