Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025781.2 | NP_080057.1 | 2 | 329 | ||
GenBank mRNA | AK007926.1 | - | 2 | 373 | ||
AK018496.1 | - | 2 | 330 | |||
BC139377.1 | - | 2 | 404 | 8862399 | ||
BC145964.1 | - | 2 | 404 | 40131267 |