Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025785.2 | NP_080061.1 | 9 | 923 | ||
GenBank mRNA | AK008027.1 | - | 9 | 927 | ||
AK018626.1 | - | 9 | 924 | |||
AK134722.1 | - | 9 | 924 | |||
AK188178.1 | - | 2 | 52 | |||
BC014749.1 | - | 9 | 909 | 4240953 |