Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029979.3 | NP_084255.2 | 6 | 1250 | ||
GenBank mRNA | AB060155.1 | - | 6 | 1194 | ||
AF145374.1 | - | 6 | 1234 | |||
AK003000.1 | - | 6 | 1209 | |||
AK032635.1 | - | 6 | 1223 | |||
AK075877.1 | - | 6 | 1221 | |||
AK146541.1 | - | 5 | 701 | |||
BC049105.1 | - | 6 | 1200 | 6418046 | ||
BC053494.1 | - | 6 | 1115 | |||
BC145783.1 | - | 6 | 1244 | 40131086 | ||
BC145809.1 | - | 6 | 1244 | 40131112 |