Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301020.1 | NP_001287949.1 | 5 | 1050 | ||
NM_004152.3 | NP_004143.1 | 5 | 1056 | |||
GenBank mRNA | AL542477.3 | - | 3 | 627 |