Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1200014J11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025818.3 | NP_080094.3 | 10 | 1308 | ||
GenBank mRNA | AK004766.1 | - | 10 | 1296 | ||
AK011450.1 | - | 10 | 1283 | |||
AK017496.1 | - | 10 | 1303 | |||
AK032983.1 | - | 8 | 1015 | |||
AK033682.1 | - | 10 | 1297 | |||
AK153152.1 | - | 10 | 1296 | |||
AK153241.1 | - | 10 | 1296 | |||
AK166563.1 | - | 8 | 1098 | |||
BC053720.1 | - | 8 | 1097 | |||
BC098495.1 | - | 10 | 1280 | 30021633 | ||
BC118943.2 | - | 10 | 1296 | 40092337 |