Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025825.3 | NP_080101.1 | 11 | 1637 | ||
GenBank mRNA | AK004888.1 | - | 11 | 1647 | ||
AK005440.1 | - | 11 | 1343 | |||
AK122222.1 | - | 1244 | ||||
AK133853.1 | - | 11 | 1629 | |||
BC018442.1 | - | 11 | 1615 | 3157290 | ||
BC019152.1 | - | 11 | 1638 | 5065139 |