Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024433.2 | NP_077753.1 | 6 | 732 | ||
GenBank mRNA | AB056100.1 | - | 6 | 720 | ||
AK005064.1 | - | 6 | 679 | |||
AK011421.1 | - | 6 | 677 | |||
AK089916.1 | - | 6 | 669 | |||
AK167319.1 | - | 6 | 733 | |||
BC003858.1 | - | 6 | 676 | 3485422 |