Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025843.3 | NP_080119.1 | 2 | 228 | ||
GenBank mRNA | AK002678.1 | - | 2 | 226 | ||
AK003300.1 | - | 2 | 229 | |||
BC034787.1 | - | 2 | 207 | 4913656 | ||
BC056984.1 | - | 1 | 4069 |