Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025860.3 | NP_080136.2 | 6 | 911 | ||
GenBank mRNA | AK019845.1 | - | 6 | 835 | ||
AK075864.1 | - | 6 | 848 | |||
BC017160.1 | - | 6 | 831 | |||
BC028246.1 | - | 6 | 882 | 2582151 | ||
BC103776.1 | - | 6 | 848 | 6492142 | ||
BC141230.1 | - | 6 | 850 | 9055842 | ||
BC141231.1 | - | 6 | 850 | 9055843 |