Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033573.1 | NP_001028745.1 | 2 | 646 | ||
NM_025883.3 | NP_080159.3 | 2 | 530 | |||
GenBank mRNA | AK010296.1 | - | 2 | 525 | ||
AK011373.1 | - | 2 | 497 | |||
AK035172.1 | - | 609 | ||||
AK038177.1 | - | 509 | ||||
AK050388.1 | - | 2 | 645 | |||
AK077693.1 | - | 2 | 484 | |||
AK077838.1 | - | 495 | ||||
AK139707.1 | - | 2 | 487 | |||
AK144128.1 | - | 2 | 502 | |||
AK154741.1 | - | 2 | 508 | |||
AK157494.1 | - | 2 | 512 | |||
AK159304.1 | - | 2 | 531 | |||
AK168497.1 | - | 2 | 525 | |||
AK181252.1 | - | 2 | 309 | |||
AK206189.1 | - | 2 | 309 | |||
AK210763.1 | - | 1 | 213 | |||
AK211713.1 | - | 1 | 311 | |||
AK217393.1 | - | 2 | 309 | |||
BC019614.2 | - | 2 | 534 | |||
BC033317.1 | - | 2 | 477 | 4459242 | ||
BX512316.1 | - | 2 | 557 |