Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024434.6 | NP_077754.3 | 2 | 260 | ||
GenBank mRNA | AF334160.1 | - | 2 | 221 | ||
AK002819.1 | - | 2 | 196 | |||
AK005334.1 | - | 2 | 173 | |||
AK008600.1 | - | 2 | 147 | |||
AK010384.1 | - | 2 | 146 | |||
AK010502.1 | - | 2 | 213 | |||
AK011604.1 | - | 2 | 146 | |||
AK089023.1 | - | 2 | 227 | |||
AK166958.1 | - | 2 | 144 | |||
AK169032.1 | - | 2 | 146 | |||
BC016536.1 | - | 2 | 260 | 3488469 | ||
CT010237.1 | - | 2 | 174 | |||
CT010301.1 | - | 2 | 174 |