Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277273.1 | NP_001264202.1 | 11, 12 | 1011 | ||
NM_025923.3 | NP_080199.1 | 11, 12 | 1026 | |||
GenBank mRNA | AF513619.1 | - | 11, 12 | 1026 | ||
AK008605.1 | - | 11, 12 | 912 | |||
AK011520.1 | - | 11, 12 | 931 | |||
AK011752.1 | - | 11, 12 | 903 | |||
AK011903.1 | - | 11, 12 | 894 | |||
AK017431.1 | - | 11, 12 | 906 | |||
AK018018.1 | - | 11, 12 | 908 | |||
AK076017.1 | - | 11, 12 | 909 | |||
AK168538.1 | - | 11, 12 | 910 | |||
BC032876.1 | - | 11, 12 | 900 | 1282316 | ||
BC066181.1 | - | 11, 12 | 955 | 6435171 |