Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025938.4 | NP_080214.1 | 5 | 507 | ||
GenBank mRNA | AK012105.1 | - | 5 | 454 | ||
AK016631.1 | - | 5 | 454 | |||
BC026988.1 | - | 5 | 437 | 5368565 | ||
BC046803.1 | - | 5 | 435 | 6404792 | ||
CF896465.1 | - | 5 | 507 |