Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012312.1 | - | 6 | 856 | ||
AK016651.1 | - | 6 | 999 | |||
AK038578.1 | - | 6 | 821 | |||
AK044291.1 | - | 6 | 969 | |||
AK048044.1 | - | 6 | 969 | |||
AK076231.1 | - | 6 | 983 | |||
AK078139.1 | - | 6 | 967 | |||
AK078468.1 | - | 6 | 983 | |||
AK078872.1 | - | 6 | 903 | |||
AK079556.1 | - | 6 | 820 | |||
AK161465.1 | - | 6 | 878 | |||
AK165398.1 | - | 6 | 985 | |||
BC031761.1 | - | 6 | 849 | 4235549 |