Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025969.4 | NP_080245.1 | 6 | 889 | ||
GenBank mRNA | AK006594.1 | - | 6 | 394 | ||
AK012203.1 | - | 6 | 889 | |||
AK013281.1 | - | 6 | 802 | |||
AK017477.1 | - | 6 | 856 | |||
AK029037.1 | - | 4 | 2066 | |||
AK032468.1 | - | 4 | 2070 | |||
AK168105.1 | - | 6 | 800 | |||
BC034297.1 | - | 6 | 864 | 4511762 |