Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164614.1 | NP_001158086.1 | 3 | 485 | ||
NM_025977.3 | NP_080253.2 | 3 | 471 | |||
GenBank mRNA | AK011114.1 | - | 3 | 472 | ||
AK016073.1 | - | 3 | 296 | |||
AK042884.1 | - | 3 | 337 | |||
AK148448.1 | - | 3 | 345 | |||
BC120702.1 | - | 3 | 446 | 40129625 | ||
BC120704.1 | - | 3 | 446 | 40129627 | ||
BC145014.1 | - | 3 | 460 |