Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025989.3 | NP_080265.2 | 8 | 1489 | ||
GenBank mRNA | AK009323.1 | - | 8 | 1490 | ||
AK009466.1 | - | 3 | 386 | |||
AK009558.1 | - | 2 | 199 | |||
AK009661.1 | - | 8 | 1490 | |||
AK009843.1 | - | 8 | 1443 | |||
BC119573.2 | - | 8 | 1484 | 40057995 |