Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026001.2 | NP_080277.1 | 11 | 868 | ||
GenBank mRNA | AK003828.1 | - | 1 | 761 | ||
AK011914.1 | - | 11 | 907 | |||
AK082752.1 | - | 11 | 869 | |||
BC047997.1 | - | 11 | 869 | 6510494 |