Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026007.4 | NP_080283.3 | 10 | 1354 | ||
GenBank mRNA | AF321126.1 | - | 9 | 1197 | ||
AK007869.1 | - | 10 | 1199 | |||
AK007991.1 | - | 10 | 1202 | |||
AK011951.1 | - | 9 | 1196 | |||
AK012431.1 | - | 10 | 1200 | |||
AK050636.1 | - | 10 | 1205 | |||
AK081859.1 | - | 10 | 1202 | |||
AK145519.1 | - | 10 | 1355 | |||
AK145565.1 | - | 10 | 1203 | |||
AK147980.1 | - | 10 | 1202 | |||
AK159644.1 | - | 10 | 1200 | |||
AK159768.1 | - | 10 | 1200 | |||
AK166765.1 | - | 10 | 1200 | |||
AK167831.1 | - | 10 | 1202 | |||
AK168749.1 | - | 10 | 1201 | |||
BC023495.1 | - | 9 | 1164 | 5251765 | ||
BC083071.1 | - | 10 | 1186 | 6329652 | ||
BC099413.1 | - | 9 | 1182 | 30839877 | ||
BC117547.1 | - | 9 | 1197 | 40105371 | ||
BC118522.1 | - | 9 | 1197 | 40105372 |