Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026029.3 | NP_080305.2 | 4, 5 | 469 | ||
GenBank mRNA | AK006979.1 | - | 4, 5 | 792 | ||
AK007439.1 | - | 4, 5 | 434 | |||
AK010992.1 | - | 4, 5 | 427 | |||
AK012569.1 | - | 4, 5 | 447 | |||
AK013212.1 | - | 4, 5 | 447 | |||
AK138073.1 | - | 446 | ||||
BC049937.1 | - | 1, 2 | 98 | |||
BC061012.1 | - | 4, 5 | 461 | 6587019 |