Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023324.2 | NP_075813.2 | 3 | 519 | ||
GenBank mRNA | AF302503.1 | - | 2 | 741 | ||
AK020915.1 | - | 3 | 553 | |||
AK029586.1 | - | 3 | 417 | |||
AK151660.1 | - | 3 | 520 | |||
AK152900.1 | - | 3 | 520 | |||
AK152997.1 | - | 3 | 520 | |||
AK153372.1 | - | 3 | 520 | |||
AK167525.1 | - | 3 | 520 | |||
BC016515.1 | - | 2 | 118 | 3600130 | ||
BC026695.1 | - | 3 | 483 |