Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145456.2 | NP_663431.2 | 14 | 3094 | ||
GenBank mRNA | AK037945.1 | - | 10 | 1689 | ||
AK122528.1 | - | 13 | 2362 | |||
BC021311.1 | - | 5 | 771 | |||
BC021521.1 | - | 5 | 974 |