Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026062.4 | NP_080338.1 | 2 | 254 | ||
GenBank mRNA | AK008504.1 | - | 2 | 197 | ||
AK013580.1 | - | 2 | 218 | |||
AK038811.1 | - | 2 | 211 | |||
AK048979.1 | - | 2 | 211 | |||
AK050305.1 | - | 2 | 211 | |||
AK053273.1 | - | 1 | 59 | |||
AK153488.1 | - | 2 | 209 | |||
BC055842.1 | - | 2 | 186 | |||
BC061013.1 | - | 2 | 206 | |||
BC095935.1 | - | 2 | 188 | |||
BC116683.1 | - | 2 | 199 | 40106565 | ||
BY131147.1 | - | 2 | 256 |