Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026062.4 | NP_080338.1 | 2 | 216 | ||
GenBank mRNA | AK008504.1 | - | 2 | 159 | ||
AK013580.1 | - | 2 | 180 | |||
AK038811.1 | - | 2 | 173 | |||
AK048979.1 | - | 2 | 173 | |||
AK050305.1 | - | 2 | 173 | |||
AK053273.1 | - | 1 | 21 | |||
AK153488.1 | - | 2 | 171 | |||
BC055842.1 | - | 2 | 148 | |||
BC061013.1 | - | 2 | 168 | |||
BC095935.1 | - | 2 | 150 | |||
BC116683.1 | - | 2 | 161 | 40106565 | ||
BY131147.1 | - | 2 | 218 |