Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110040N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026077.3 | NP_080353.1 | 2 | 163 | ||
GenBank mRNA | AK014163.1 | - | 2 | 143 | ||
AK018003.1 | - | 2 | 189 | |||
AK019261.1 | - | 2 | 163 | |||
AK170168.1 | - | 2 | 180 | |||
BC027500.1 | - | 2 | 151 | 1448876 |