Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000528.3 | NP_000519.2 | 9 | 1242 | ||
NM_001173498.1 | NP_001166969.1 | 9 | 1239 | |||
XM_005259913.1 | XP_005259970.1 | 9 | 1223 | |||
XM_017026818.1 | XP_016882307.1 | 6 | 821 | |||
GenBank mRNA | AB209921.1 | - | 9 | 1154 | ||
AK291572.1 | - | 9 | 1207 | |||
AK303418.1 | - | 10 | 1181 | |||
BC000736.2 | - | 9 | 1188 | 3504417 | ||
U05572.1 | - | 9 | 1196 | |||
U60266.1 | - | 9 | 1180 | |||
U68382.1 | - | 8 | 1064 | |||
U68567.1 | - | 9 | 1154 |