Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700037H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026091.2 | NP_080367.1 | 6 | 624 | ||
GenBank mRNA | AK003406.1 | - | 6 | 625 | ||
AK040365.1 | - | 3 | 1030 | |||
BC006041.1 | - | 5 | 536 | 3497476 | ||
BC071212.1 | - | 7 | 740 |