Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8504
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
11
1500
NM_002374.3
NP_002365.3
12
5194
NM_031845.2
NP_114033.2
9
1126
NM_031847.2
NP_114035.2
9
1126
XM_005246555.1
XP_005246612.1
14
5730
XM_005246565.1
XP_005246622.1
10
5019
XM_005246566.1
XP_005246623.1
10
5007
XM_006712532.1
XP_006712595.1
14
5727
XM_011511186.1
XP_011509488.1
14
5637
XM_011511187.1
XP_011509489.1
13
5531
XM_011511188.1
XP_011509490.1
13
5531
XM_011511190.1
XP_011509492.1
13
5534
XM_011511191.1
XP_011509493.1
12
5399
XM_011511192.1
XP_011509494.1
12
5363
XM_011511193.1
XP_011509495.1
12
5360
XM_011511195.1
XP_011509497.1
12
5285
XM_011511196.2
XP_011509498.1
13
5472
XM_011511197.2
XP_011509499.1
13
5469
XM_017004111.1
XP_016859600.1
12
5360
XM_017004112.1
XP_016859601.1
13
5484
XM_017004113.1
XP_016859602.1
13
5376
XM_017004114.1
XP_016859603.1
12
5313
XM_017004115.1
XP_016859604.1
11
5105
XM_017004116.1
XP_016859605.1
11
5102
XM_017004117.1
XP_016859606.1
11
5102
XM_017004118.1
XP_016859607.1
12
5220
XM_017004119.1
XP_016859608.1
12
5310
XM_017004120.1
XP_016859609.1
12
5310
XM_017004121.1
XP_016859610.1
12
5217
XM_017004122.1
XP_016859611.1
12
5217
XM_017004123.1
XP_016859612.1
12
5301
XM_017004124.1
XP_016859613.1
12
5208
XM_017004125.1
XP_016859614.1
12
5298
XM_017004126.1
XP_016859615.1
12
5298
XM_017004127.1
XP_016859616.1
12
5298
XM_017004128.1
XP_016859617.1
11
5099
XM_017004129.1
XP_016859618.1
11
5096
XM_017004130.1
XP_016859619.1
10
5456
XM_017004131.1
XP_016859620.1
9
4848
XM_017004132.1
XP_016859621.1
9
4836
XM_017004133.1
XP_016859622.1
9
4848
XM_017004134.1
XP_016859623.1
9
4836
XM_017004135.1
XP_016859624.1
9
5282
XM_017004136.1
XP_016859625.1
9
4845
XM_017004137.1
XP_016859626.1
9
4833
XM_017004138.1
XP_016859627.1
10
1323
XM_017004139.1
XP_016859628.1
7
1080
XM_017004140.1
XP_016859629.1
9
1152
GenBank mRNA
AB209330.1
-
8
1044
AK291446.1
-
6
915
AL833567.1
-
3
611
BC027583.1
-
9
1198
BC038857.1
-
11
1489
BC110423.1
-
9
1374
BC117123.1
-
7
955
BC143245.1
-
7
960
S76756.1
-
2
267
U01828.1
-
12
5194
U89329.1
-
6
878
U89330.1
-
9
4934
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0