Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026225.3 | NP_080501.2 | 3 | 381 | ||
GenBank mRNA | AB093286.1 | - | 3 | 353 | ||
AK016663.1 | - | 3 | 394 | |||
AK031149.1 | - | 3 | 375 | |||
AK045223.1 | - | 3 | 375 | |||
AK048986.1 | - | 3 | 368 | |||
AK153320.1 | - | 3 | 370 | |||
BC025427.1 | - | 3 | 387 | 2649589 |