Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024203.3 | NP_077165.2 | 2 | 948 | ||
GenBank mRNA | AK016568.1 | - | 2 | 916 | ||
AK028911.1 | - | 2 | 878 | |||
AK050319.1 | - | 1 | 663 | |||
AK076185.1 | - | 2 | 914 | |||
AK078215.1 | - | 2 | 945 | |||
AK087438.1 | - | 2 | 894 | |||
AK132211.1 | - | 2 | 926 | |||
AK220425.1 | - | 2 | 910 | |||
BC066088.1 | - | 2 | 992 | |||
BC085163.1 | - | 2 | 946 | 30618952 | ||
DQ873694.1 | - | 2 | 929 |