Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026331.3 | NP_080607.2 | 4 | 1080 | ||
GenBank mRNA | AF288621.1 | - | 4 | 1033 | ||
AF361699.1 | - | 3 | 2790 | |||
AK006155.1 | - | 4 | 1095 | |||
AK015770.1 | - | 4 | 780 | |||
AK028588.1 | - | 2 | 2147 | |||
AK046008.1 | - | 4 | 1458 | |||
AK050414.1 | - | 4 | 1048 | |||
AK162615.1 | - | 2 | 942 | |||
BC040399.1 | - | 2 | 903 | 4037680 |