Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026370.1 | NP_080646.1 | 10 | 1228 | ||
GenBank mRNA | AK003264.1 | - | 10 | 1228 | ||
AK008418.1 | - | 10 | 1215 | |||
AK017161.1 | - | 9 | 974 | |||
AK018002.1 | - | 10 | 1224 | |||
AK132000.1 | - | 10 | 1229 | |||
AK146710.1 | - | 10 | 1230 | |||
AK209737.1 | - | 2 | 96 | |||
BC036284.1 | - | 10 | 1236 | 5136304 |