Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026388.2 | NP_080664.1 | 2 | 392 | ||
GenBank mRNA | AK004777.1 | - | 2 | 375 | ||
AK155413.1 | - | 2 | 387 | |||
AK219916.1 | - | 2 | 135 | |||
BC019544.1 | - | 2 | 356 | 4211201 | ||
BY739996.1 | - | 2 | 392 |