Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026395.1 | NP_080671.1 | 6 | 590 | ||
GenBank mRNA | AK002550.1 | - | 6 | 590 | ||
AK003682.1 | - | 6 | 612 | |||
AK013262.1 | - | 6 | 647 | |||
AK078396.1 | - | 6 | 653 | |||
AK168192.1 | - | 7 | 809 | |||
BC029189.1 | - | 5 | 496 | 4036721 |