Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026403.3 | NP_080679.3 | 5 | 1177 | ||
GenBank mRNA | AK011570.1 | - | 5 | 1167 | ||
AK015585.1 | - | 5 | 927 | |||
AK032873.1 | - | 5 | 1161 | |||
AK037831.1 | - | 5 | 1165 | |||
AK050912.1 | - | 5 | 1168 | |||
AK083861.1 | - | 5 | 1046 | |||
AK139689.1 | - | 5 | 1128 | |||
AK146020.1 | - | 5 | 1132 | |||
AK146687.1 | - | 5 | 1149 | |||
AK152732.1 | - | 5 | 1151 | |||
AK154387.1 | - | 5 | 1169 | |||
AK159798.1 | - | 5 | 1151 | |||
AK178392.1 | - | 2 | 297 | |||
BC018523.1 | - | 5 | 1157 | 3499986 |