Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026419.2 | NP_080695.1 | 5 | 461 | ||
GenBank mRNA | AK008858.1 | - | 5 | 460 | ||
AK009129.1 | - | 5 | 460 | |||
AK010149.1 | - | 5 | 461 | |||
BC056210.1 | - | 4 | 400 | |||
BC061066.1 | - | 5 | 461 | 30309250 |
Gene Symbol: Cela3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126318.1 | NP_001119790.1 | 5 | 472 | ||
GenBank mRNA | BM501883.1 | - | 412 |