Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018771.3 | NP_061241.1 | 7 | 977 | ||
GenBank mRNA | AF061263.1 | - | 6 | 911 | ||
AF104358.1 | - | 7 | 955 | |||
AK002492.1 | - | 7 | 957 | |||
AK086506.1 | - | 7 | 978 | |||
BC003490.1 | - | 7 | 977 | 2811749 |