Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026456.4 | NP_080732.1 | 3 | 147 | ||
GenBank mRNA | AK011757.1 | - | 3 | 155 | ||
AK011958.1 | - | 3 | 167 | |||
AK012909.1 | - | 3 | 148 | |||
AK013089.1 | - | 3 | 164 | |||
AK039746.1 | - | 3 | 148 | |||
BC009104.1 | - | 3 | 147 | 3483215 | ||
BC028545.1 | - | 3 | 129 | 1399237 |