Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026469.2 | NP_080745.2 | 2 | 896 | ||
GenBank mRNA | AK006327.1 | - | 1 | 176 |