Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026474.5 | NP_080750.1 | 12 | 897 | ||
GenBank mRNA | AK007881.1 | - | 12 | 832 | ||
AK010711.1 | - | 11 | 866 | |||
AK010830.1 | - | 12 | 814 | |||
AK020433.1 | - | 12 | 816 | |||
AK045153.1 | - | 12 | 836 | |||
AK082328.1 | - | 12 | 899 | |||
AK141416.1 | - | 833 | ||||
AK148838.1 | - | 12 | 811 | |||
BC009167.1 | - | 12 | 814 | 2811924 | ||
BC038677.1 | - | 295 |