Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026474.5 | NP_080750.1 | 4 | 337 | ||
GenBank mRNA | AK007881.1 | - | 4 | 272 | ||
AK010711.1 | - | 3 | 306 | |||
AK010830.1 | - | 4 | 254 | |||
AK020433.1 | - | 4 | 256 | |||
AK045153.1 | - | 4 | 276 | |||
AK082328.1 | - | 4 | 339 | |||
AK141416.1 | - | 273 | ||||
AK148838.1 | - | 4 | 251 | |||
BC009167.1 | - | 4 | 254 | 2811924 |