Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114977.1 | NP_001108449.1 | 12 | 1333 | ||
NM_026476.2 | NP_080752.2 | 11 | 1201 | |||
GenBank mRNA | AK029898.1 | - | 12 | 1233 | ||
AK179363.1 | - | 1 | 62 | |||
BC031497.1 | - | 12 | 1193 | |||
BC068265.1 | - | 11 | 1217 | 30022109 | ||
BC080699.1 | - | 11 | 1082 |