Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023409.4 | NP_075898.1 | 1, 2 | 336 | ||
GenBank mRNA | AB021289.1 | - | 1, 2 | 91 | ||
AI391386.1 | - | 1, 2 | 336 | |||
AK008603.1 | - | 1, 2 | 123 | |||
AK009127.1 | - | 1, 2 | 153 | |||
AK012239.1 | - | 1, 2 | 183 | |||
AK149805.1 | - | 1, 2 | 125 | |||
AK151067.1 | - | 1, 2 | 123 | |||
AK151133.1 | - | 1, 2 | 124 | |||
AK152236.1 | - | 1, 2 | 123 | |||
AK153020.1 | - | 1, 2 | 123 | |||
AK153203.1 | - | 1, 2 | 123 | |||
AK154838.1 | - | 1, 2 | 123 | |||
AK158624.1 | - | 1, 2 | 304 | |||
AK159154.1 | - | 1, 2 | 170 | |||
AK165263.1 | - | 1, 2 | 149 | |||
AK167869.1 | - | 1, 2 | 124 | |||
AK170327.1 | - | 1, 2 | 124 | |||
BC003471.1 | - | 1, 2 | 138 | 2649564 | ||
BC007190.1 | - | 1, 2 | 170 | 3592515 |